Search
Search

UCKL1
UCKL1-AS1
ZNF512BCCCACCTTCCGGCCGGTGGAGCCCC[A/G]GGCCCCCTTGTCTCTGCATCCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
UCKL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| UCKL1-AS1 - UCKL1 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ZNF512B - zinc finger protein 512B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020713.2 | 2760 | Missense Mutation | CGG,TGG | R,W 877 | NP_065764.1 | |
| XM_005260226.3 | 2760 | Missense Mutation | CGG,TGG | R,W 891 | XP_005260283.1 | |
| XM_011528929.2 | 2760 | Missense Mutation | CGG,TGG | R,W 877 | XP_011527231.1 | |
| XM_011528930.2 | 2760 | Missense Mutation | CGG,TGG | R,W 858 | XP_011527232.1 | |
| XM_011528931.2 | 2760 | Missense Mutation | CGG,TGG | R,W 547 | XP_011527233.1 | |
| XM_011528932.2 | 2760 | Missense Mutation | CGG,TGG | R,W 546 | XP_011527234.1 | |
| XM_011528933.2 | 2760 | Intron | XP_011527235.1 | |||