Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GCTATCTGGGAGGGCTAAACAGCCT[C/G]GAGGCTGAGTCTGCAGAGGAAGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615884 MIM: 603253 | ||||||||||||||||||||
Literature Links: |
APMAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APMAP - adipocyte plasma membrane associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020531.2 | 1341 | Missense Mutation | CAG,GAG | Q,E 414 | NP_065392.1 | |
XM_005260763.3 | 1341 | UTR 3 | XP_005260820.1 |
CST7 - cystatin F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |