Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTCTCAGATCGGTTAATGCAG[C/T]TGAATTTTTGCTTGAGAATGTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610952 MIM: 610833 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRNKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRNKL1 - crooked neck pre-mRNA splicing factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278625.1 | 3749 | UTR 3 | NP_001265554.1 | |||
NM_001278626.1 | 3749 | UTR 3 | NP_001265555.1 | |||
NM_001278627.1 | 3749 | UTR 3 | NP_001265556.1 | |||
NM_001278628.1 | 3749 | UTR 3 | NP_001265557.1 | |||
NM_016652.5 | 3749 | UTR 3 | NP_057736.4 |
NAA20 - N(alpha)-acetyltransferase 20, NatB catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |