Search Thermo Fisher Scientific
GCAACAGTAGCACATCTGCGGGGAA[C/T]TTTCTCAGGTACCATTTTTGCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605296 | ||||||||||||||||||||
Literature Links: |
BRWD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRWD1 - bromodomain and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007246.2 | 6983 | Intron | NP_001007247.1 | |||
NM_018963.4 | 6983 | Missense Mutation | ATT,GTT | I,V 2230 | NP_061836.2 | |
NM_033656.3 | 6983 | UTR 3 | NP_387505.1 | |||
XM_011529611.1 | 6983 | Missense Mutation | ATT,GTT | I,V 2182 | XP_011527913.1 | |
XM_011529612.2 | 6983 | Missense Mutation | ATT,GTT | I,V 1950 | XP_011527914.1 | |
XM_011529613.1 | 6983 | Missense Mutation | ATT,GTT | I,V 1241 | XP_011527915.1 | |
XM_017028373.1 | 6983 | Missense Mutation | ATT,GTT | I,V 2143 | XP_016883862.1 | |
XM_017028374.1 | 6983 | Missense Mutation | ATT,GTT | I,V 1241 | XP_016883863.1 | |
XM_017028375.1 | 6983 | Intron | XP_016883864.1 |
PSMG1 - proteasome assembly chaperone 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |