Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGGAGGAGGTACGACTCAGGCGAGG[C/T]AGTGAAGAATGACTGCGTAGTAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611845 | ||||||||||||||||||||
Literature Links: |
LINC00515 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00515 - long intergenic non-protein coding RNA 515 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL39 - mitochondrial ribosomal protein L39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017446.3 | 1024 | Intron | NP_059142.2 | |||
NM_080794.3 | 1024 | Missense Mutation | ACC,GCC | T,A 340 | NP_542984.2 | |
XM_006724026.3 | 1024 | Missense Mutation | ACC,GCC | T,A 340 | XP_006724089.1 | |
XM_011529651.2 | 1024 | Intron | XP_011527953.1 |