Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCCTGAGACACTATGGCCCTGA[C/T]CTCAGACCTGGGGAAACAGATAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602385 | ||||||||||||||||||||
Literature Links: |
SULT1C2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SULT1C2 - sulfotransferase family 1C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001056.3 | 464 | Missense Mutation | ACC,ATC | T,I 4 | NP_001047.1 | |
NM_176825.2 | 464 | Missense Mutation | ACC,ATC | T,I 4 | NP_789795.1 | |
XM_005264013.3 | 464 | Missense Mutation | ACC,ATC | T,I 4 | XP_005264070.1 |