Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AAAATAGATTATCACTCACTTGTAT[C/G]GTCTTCATATCGTTCTATATAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611972 | ||||||||||||||||||||
Literature Links: |
MRPS5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPS5 - mitochondrial ribosomal protein S5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321995.1 | 919 | Missense Mutation | CAT,GAT | H,D 209 | NP_001308924.1 | |
NM_001321996.1 | 919 | Missense Mutation | CAT,GAT | H,D 122 | NP_001308925.1 | |
NM_001321997.1 | 919 | Missense Mutation | CAT,GAT | H,D 122 | NP_001308926.1 | |
NM_031902.4 | 919 | Missense Mutation | CAT,GAT | H,D 288 | NP_114108.1 |