Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTAAAGCAGGCAGTTTCTTGCT[C/T]ATGCCACACGATATGCCTTCATTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CLDND1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDND1 - claudin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040181.1 | 687 | Silent Mutation | TAA,TGA | *,* 254 | NP_001035271.1 | |
NM_001040182.1 | 687 | Silent Mutation | TAA,TGA | *,* 277 | NP_001035272.1 | |
NM_001040183.1 | 687 | Silent Mutation | TAA,TGA | *,* 254 | NP_001035273.1 | |
NM_001040199.1 | 687 | Silent Mutation | TAA,TGA | *,* 254 | NP_001035289.1 | |
NM_001040200.1 | 687 | Silent Mutation | TAA,TGA | *,* 159 | NP_001035290.1 | |
NM_019895.2 | 687 | Silent Mutation | TAA,TGA | *,* 254 | NP_063948.1 |