Search
Search

GRIA1TTCCTTCAAAAACTGTATTCTGTGG[C/T]GGCTAGTGGCTGCTGCTTTGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| GRIA1 - glutamate ionotropic receptor AMPA type subunit 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000827.3 | 19 | Intron | NP_000818.2 | |||
| NM_001114183.1 | 19 | Intron | NP_001107655.1 | |||
| NM_001258019.1 | 19 | Intron | NP_001244948.1 | |||
| NM_001258020.1 | 19 | Intron | NP_001244949.1 | |||
| NM_001258021.1 | 19 | Intron | NP_001244950.1 | |||
| NM_001258022.1 | 19 | Intron | NP_001244951.1 | |||
| NM_001258023.1 | 19 | Intron | NP_001244952.1 | |||
| XM_011537635.2 | 19 | UTR 5 | XP_011535937.1 | |||
| XM_017009392.1 | 19 | Intron | XP_016864881.1 | |||
| XM_017009393.1 | 19 | Intron | XP_016864882.1 | |||