Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCGTGTTTACAAGTATGTACACA[A/C]AAAGTCAGGGTTCAGGAGGCCGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRY4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRY4 - sprouty RTK signaling antagonist 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127496.1 | 3556 | UTR 3 | NP_001120968.1 | |||
NM_001293289.1 | 3556 | UTR 3 | NP_001280218.1 | |||
NM_001293290.1 | 3556 | UTR 3 | NP_001280219.1 | |||
NM_030964.3 | 3556 | UTR 3 | NP_112226.2 | |||
XM_011537685.2 | 3556 | UTR 3 | XP_011535987.1 | |||
XM_017009910.1 | 3556 | UTR 3 | XP_016865399.1 |
SPRY4-IT1 - SPRY4 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |