Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAAGTAAACTTGAGCTGGTAC[A/G]ATGGAACGGTCAACCCCTGCCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SPINK14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPINK14 - serine peptidase inhibitor, Kazal type 14 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001325.2 | 318 | Missense Mutation | AAT,GAT | N,D 51 | NP_001001325.1 | |
XM_017009466.1 | 318 | Missense Mutation | AAT,GAT | N,D 68 | XP_016864955.1 | |
XM_017009467.1 | 318 | Intron | XP_016864956.1 | |||
XM_017009468.1 | 318 | Missense Mutation | AAT,GAT | N,D 44 | XP_016864957.1 | |
XM_017009469.1 | 318 | Missense Mutation | AAT,GAT | N,D 51 | XP_016864958.1 |