Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GTCGAGAAGTCCGGAAGCTTGTGGC[A/G]CTGAAAACGAACGGTAAGACTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191163 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130476 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100130476 - uncharacterized LOC100130476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107986649 - uncharacterized LOC107986649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP3 - TNF alpha induced protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270507.1 | 97 | Silent Mutation | GCA,GCG | A,A 94 | NP_001257436.1 | |
NM_001270508.1 | 97 | Silent Mutation | GCA,GCG | A,A 94 | NP_001257437.1 | |
NM_006290.3 | 97 | Silent Mutation | GCA,GCG | A,A 94 | NP_006281.1 | |
XM_005267119.1 | 97 | Silent Mutation | GCA,GCG | A,A 94 | XP_005267176.1 | |
XM_006715555.1 | 97 | UTR 5 | XP_006715618.1 | |||
XM_011536095.1 | 97 | Silent Mutation | GCA,GCG | A,A 94 | XP_011534397.1 | |
XM_011536096.1 | 97 | Silent Mutation | GCA,GCG | A,A 94 | XP_011534398.1 |