Search
Search

HIST1H2BH
HIST1H3F
HIST1H4F
HIST1H4GGTCACTCGCTTGGCGTGGATAGCAC[A/C]CAGGTTGGTGTCCTCAAAGAGCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
HIST1H2BH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| HIST1H2BH - histone cluster 1, H2bh | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HIST1H3F - histone cluster 1, H3f | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_021018.2 | Intron | NP_066298.1 | ||||
| HIST1H4F - histone cluster 1, H4f | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HIST1H4G - histone cluster 1, H4g | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||