Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AAGAAGGACAAGAAGATCAAAAAAA[C/T]GGTGAGAAAATGAGGGTTGAGGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603429 MIM: 611619 | ||||||||||||||||||||
Literature Links: |
ABCF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCF1 - ATP binding cassette subfamily F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025091.1 | 214 | Missense Mutation | ACG,ATG | T,M 40 | NP_001020262.1 | |
NM_001090.2 | 214 | Missense Mutation | ACG,ATG | T,M 40 | NP_001081.1 |
MIR877 - microRNA 877 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |