Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CAGAGAAAATTGTGATATCAATAGC[A/G]TTGTAACTCAGAATGGTGAAGTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615532 MIM: 186977 | ||||||||||||||||||||
Literature Links: |
ERMARD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERMARD - ER membrane associated RNA degradation | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278531.1 | 1235 | Missense Mutation | ATT,GTT | I,V 37 | NP_001265460.1 | |
NM_001278532.1 | 1235 | Intron | NP_001265461.1 | |||
NM_001278533.1 | 1235 | Missense Mutation | ATT,GTT | I,V 37 | NP_001265462.1 | |
NM_018341.2 | 1235 | Missense Mutation | ATT,GTT | I,V 37 | NP_060811.1 | |
XM_011535938.2 | 1235 | Missense Mutation | ATT,GTT | I,V 37 | XP_011534240.1 | |
XM_011535939.2 | 1235 | UTR 5 | XP_011534241.1 | |||
XM_011535940.2 | 1235 | UTR 5 | XP_011534242.1 | |||
XM_017011027.1 | 1235 | UTR 5 | XP_016866516.1 | |||
XM_017011028.1 | 1235 | UTR 5 | XP_016866517.1 | |||
XM_017011029.1 | 1235 | UTR 5 | XP_016866518.1 | |||
XM_017011030.1 | 1235 | Intron | XP_016866519.1 | |||
XM_017011031.1 | 1235 | UTR 5 | XP_016866520.1 | |||
XM_017011032.1 | 1235 | UTR 5 | XP_016866521.1 | |||
XM_017011033.1 | 1235 | UTR 5 | XP_016866522.1 |
TCTE3 - t-complex-associated-testis-expressed 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |