Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACATCTGAAACCTTAAACTCAC[G/T]AATACCCCACTCTCTGGCCACATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPYS - dihydropyrimidinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001385.2 | Intron | NP_001376.1 | ||||
XM_005250818.3 | Intron | XP_005250875.1 | ||||
XM_006716518.3 | Intron | XP_006716581.1 | ||||
XM_011516903.2 | Intron | XP_011515205.1 | ||||
XM_017013167.1 | Intron | XP_016868656.1 |