Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ACTGAGAGTGGAGCGCTTGCCGCTG[C/G]ATGCATACTCATCTCCAAAGGTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604912 | ||||||||||||||||||||
Literature Links: |
TAF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAF2 - TATA-box binding protein associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003184.3 | 3509 | Missense Mutation | CCA,GCA | P,A 1125 | NP_003175.1 | |
XM_006716621.3 | 3509 | Missense Mutation | CCA,GCA | P,A 1177 | XP_006716684.1 | |
XM_011517259.2 | 3509 | Missense Mutation | CCA,GCA | P,A 1140 | XP_011515561.1 | |
XM_017013779.1 | 3509 | Missense Mutation | CCA,GCA | P,A 1088 | XP_016869268.1 | |
XM_017013780.1 | 3509 | Missense Mutation | CCA,GCA | P,A 1008 | XP_016869269.1 | |
XM_017013781.1 | 3509 | Missense Mutation | CCA,GCA | P,A 1008 | XP_016869270.1 | |
XM_017013782.1 | 3509 | Missense Mutation | CCA,GCA | P,A 1008 | XP_016869271.1 | |
XM_017013783.1 | 3509 | Missense Mutation | CCA,GCA | P,A 966 | XP_016869272.1 | |
XM_017013784.1 | 3509 | Missense Mutation | CCA,GCA | P,A 956 | XP_016869273.1 | |
XM_017013785.1 | 3509 | Missense Mutation | CCA,GCA | P,A 956 | XP_016869274.1 | |
XM_017013786.1 | 3509 | Missense Mutation | CCA,GCA | P,A 956 | XP_016869275.1 | |
XM_017013787.1 | 3509 | Missense Mutation | CCA,GCA | P,A 919 | XP_016869276.1 | |
XM_017013788.1 | 3509 | Missense Mutation | CCA,GCA | P,A 760 | XP_016869277.1 | |
XM_017013789.1 | 3509 | Missense Mutation | CCA,GCA | P,A 708 | XP_016869278.1 | |
XM_017013790.1 | 3509 | Missense Mutation | CCA,GCA | P,A 708 | XP_016869279.1 | |
XM_017013791.1 | 3509 | Missense Mutation | CCA,GCA | P,A 678 | XP_016869280.1 |