Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGCAGGAGAGTTTCGGCTGCGT[A/G]GTGGCCAACCGCTTCCATCAGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HABP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HABP4 - hyaluronan binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014282.3 | 194 | Silent Mutation | GTA,GTG | V,V 23 | NP_055097.2 | |
XM_005251812.3 | 194 | Intron | XP_005251869.1 | |||
XM_017014484.1 | 194 | Intron | XP_016869973.1 |