Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGTTCCAGATGAATCCAGAGTGGGG[A/G]CAGGCCTTCGTGCACGTGGCCGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3202-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3202-1 - microRNA 3202-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3202-2 - microRNA 3202-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM187 - transmembrane protein 187 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003492.2 | 567 | Silent Mutation | GGA,GGG | G,G 6 | NP_003483.1 | |
XM_011531199.2 | 567 | Silent Mutation | GGA,GGG | G,G 6 | XP_011529501.1 |