Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGTGGGAGGTAGCTATTGTAGAAAA[C/T]TCATTGTCCCACATTGGTCTCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300907 MIM: 300225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSTF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSTF2 - cleavage stimulation factor subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOX1 - NADPH oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271815.1 | 1598 | Silent Mutation | GAA,GAG | E,E 476 | NP_001258744.1 | |
NM_007052.4 | 1598 | Silent Mutation | GAA,GAG | E,E 513 | NP_008983.2 | |
NM_013955.2 | 1598 | Silent Mutation | GAA,GAG | E,E 464 | NP_039249.1 | |
XM_017029407.1 | 1598 | Silent Mutation | GAA,GAG | E,E 407 | XP_016884896.1 |