Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCAGCCGCTTGCGAATCCATTC[C/T]ATCAGGGCCACATGCTCCACGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300092 MIM: 300044 | ||||||||||||||||||||
Literature Links: |
TEX28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX28 - testis expressed 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205201.1 | 291 | Intron | NP_001192130.1 | |||
NM_001586.2 | 291 | Intron | NP_001577.1 | |||
XM_011531116.1 | 291 | Intron | XP_011529418.1 | |||
XM_011531117.1 | 291 | Intron | XP_011529419.1 | |||
XM_011531118.2 | 291 | Intron | XP_011529420.1 |
TKTL1 - transketolase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145933.1 | 291 | Silent Mutation | TCC,TCT | S,S 35 | NP_001139405.1 | |
NM_001145934.1 | 291 | Intron | NP_001139406.1 | |||
NM_012253.3 | 291 | Silent Mutation | TCC,TCT | S,S 35 | NP_036385.3 |