Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GACACACATGCATTTTTCATTCTGT[C/T]AATTGAATTTTTCAGCTCTAAGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300654 | ||||||||||||||||||||
Literature Links: |
FAAH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAAH2 - fatty acid amide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174912.3 | Intron | NP_777572.2 | ||||
XM_005261982.1 | Intron | XP_005262039.1 | ||||
XM_005261985.2 | Intron | XP_005262042.1 | ||||
XM_005261986.3 | Intron | XP_005262043.1 | ||||
XM_005261987.3 | Intron | XP_005262044.1 | ||||
XM_011530767.2 | Intron | XP_011529069.1 | ||||
XM_017029290.1 | Intron | XP_016884779.1 | ||||
XM_017029291.1 | Intron | XP_016884780.1 | ||||
XM_017029292.1 | Intron | XP_016884781.1 | ||||
XM_017029293.1 | Intron | XP_016884782.1 | ||||
XM_017029294.1 | Intron | XP_016884783.1 |