Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGGCTGCATCTTCCTGGAAGATTGG[G/T]GGACTGACTTCACCAGCCTTGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAI2 - retinoic acid induced 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172732.1 | 1647 | Silent Mutation | CCA,CCC | P,P 284 | NP_001166203.1 | |
NM_001172739.1 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | NP_001166210.1 | |
NM_001172743.1 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | NP_001166214.1 | |
NM_021785.4 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | NP_068557.3 | |
XM_006724459.2 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | XP_006724522.1 | |
XM_006724460.1 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | XP_006724523.1 | |
XM_011545439.2 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | XP_011543741.1 | |
XM_011545440.2 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | XP_011543742.1 | |
XM_011545441.2 | 1647 | Silent Mutation | CCA,CCC | P,P 334 | XP_011543743.1 |