Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCAATGAGAGTCGGGAACACACCCA[C/T]CAGCTCCACCACGGTGATGTGTCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300187 | ||||||||||||||||||||
Literature Links: |
SRPX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRPX - sushi repeat containing protein, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170750.1 | 1209 | Missense Mutation | ATG,GTG | M,V 360 | NP_001164221.1 | |
NM_001170751.1 | 1209 | Missense Mutation | ATG,GTG | M,V 321 | NP_001164222.1 | |
NM_001170752.1 | 1209 | Intron | NP_001164223.1 | |||
NM_006307.4 | 1209 | Missense Mutation | ATG,GTG | M,V 380 | NP_006298.1 | |
XM_017029893.1 | 1209 | Missense Mutation | ATG,GTG | M,V 380 | XP_016885382.1 |