Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
TGAGTGGGTAGTAGGGACTGTTGCT[C/T]AGGATCAGGGCCACCTCACTCTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300256 MIM: 300040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B10 - hydroxysteroid 17-beta dehydrogenase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIBC1 - RIB43A domain with coiled-coils 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031745.4 | Intron | NP_001026915.1 | ||||
NM_001267053.3 | Intron | NP_001253982.1 | ||||
NM_144968.3 | Intron | NP_659405.1 | ||||
XM_005261988.3 | Intron | XP_005262045.1 | ||||
XM_005261990.3 | Intron | XP_005262047.1 |
SMC1A - structural maintenance of chromosomes 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |