Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TTAAGATATTTTATGATGTTCATAT[A/C]CTAGGAATTGTTTCTGAAGGAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS6L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INTS6L - integrator complex subunit 6 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182540.4 | Intron | NP_872346.3 | ||||
XM_005262394.2 | Intron | XP_005262451.1 | ||||
XM_006724740.2 | Intron | XP_006724803.1 | ||||
XM_006724741.3 | Intron | XP_006724804.1 | ||||
XM_011531311.2 | Intron | XP_011529613.1 | ||||
XM_017029342.1 | Intron | XP_016884831.1 | ||||
XM_017029343.1 | Intron | XP_016884832.1 | ||||
XM_017029344.1 | Intron | XP_016884833.1 | ||||
XM_017029345.1 | Intron | XP_016884834.1 | ||||
XM_017029346.1 | Intron | XP_016884835.1 | ||||
XM_017029347.1 | Intron | XP_016884836.1 | ||||
XM_017029348.1 | Intron | XP_016884837.1 | ||||
XM_017029349.1 | Intron | XP_016884838.1 |
INTS6L-AS1 - INTS6L antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |