Search
Search

AMELX
ARHGAP6CTTGGTTCTAACCCAGCTAGTAAAA[C/T]GTAAGGATTAGGTAAGATGTTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMELX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| AMELX - amelogenin, X-linked | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001142.2 | Intron | NP_001133.1 | ||||
| NM_182680.1 | Intron | NP_872621.1 | ||||
| NM_182681.1 | Intron | NP_872622.1 | ||||
| XM_017029404.1 | Intron | XP_016884893.1 | ||||
| ARHGAP6 - Rho GTPase activating protein 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001287242.1 | Intron | NP_001274171.1 | ||||
| NM_006125.2 | Intron | NP_006116.2 | ||||
| NM_013423.2 | Intron | NP_038267.1 | ||||
| NM_013427.2 | Intron | NP_038286.2 | ||||