Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TTCCCGACTCTTAAACTTCAAAACC[A/G]CATAGTTATATGTAGTAGCCATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300051 MIM: 300170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPM6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPM6B - glycoprotein M6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001994.2 | 1207 | Missense Mutation | GCG,GTG | A,V 232 | NP_001001994.1 | |
NM_001001995.2 | 1207 | Intron | NP_001001995.1 | |||
NM_001001996.2 | 1207 | Missense Mutation | GCG,GTG | A,V 291 | NP_001001996.1 | |
NM_001318729.1 | 1207 | Intron | NP_001305658.1 | |||
NM_005278.4 | 1207 | Missense Mutation | GCG,GTG | A,V 251 | NP_005269.1 | |
XM_005274489.4 | 1207 | Intron | XP_005274546.1 | |||
XM_011545497.2 | 1207 | Intron | XP_011543799.1 | |||
XM_017029432.1 | 1207 | Missense Mutation | GCG,GTG | A,V 272 | XP_016884921.1 | |
XM_017029433.1 | 1207 | Intron | XP_016884922.1 | |||
XM_017029434.1 | 1207 | Missense Mutation | GCG,GTG | A,V 165 | XP_016884923.1 |
OFD1 - OFD1, centriole and centriolar satellite protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003611.2 | 1207 | Intron | NP_003602.1 |