Search
Search

ARMCX4
GLA
HNRNPH2
RPL36A-HNRNPH2GGGGCCTACCCTGGTCCTGCTCAGC[C/T]GATGAAGTGATGCGCTTCTTCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ARMCX4 - armadillo repeat containing, X-linked 4 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| GLA - galactosidase alpha | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HNRNPH2 - heterogeneous nuclear ribonucleoprotein H2 (H') | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001032393.2 | 456 | Intron | NP_001027565.1 | |||
| NM_019597.4 | 456 | Intron | NP_062543.1 | |||
| RPL36A-HNRNPH2 - RPL36A-HNRNPH2 readthrough | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001199973.1 | 456 | UTR 3 | NP_001186902.1 | |||
| NM_001199974.1 | 456 | UTR 3 | NP_001186903.1 | |||