Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGACTGAACCAGCTGGTGGTGG[C/T]CCGATGGCTGGAGACACCCACTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM8 - lysine demethylase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145348.1 | 188 | Silent Mutation | GGC,GGT | G,G 37 | NP_001138820.1 | |
NM_024773.2 | 188 | UTR 5 | NP_079049.2 | |||
XM_017023676.1 | 188 | UTR 5 | XP_016879165.1 | |||
XM_017023677.1 | 188 | UTR 5 | XP_016879166.1 | |||
XM_017023678.1 | 188 | UTR 5 | XP_016879167.1 | |||
XM_017023679.1 | 188 | UTR 5 | XP_016879168.1 | |||
XM_017023680.1 | 188 | Intron | XP_016879169.1 |