Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TAATGCCGGAACAGCTTCAGCTTGG[A/C]CAGGTTCACTGCCACTGAGGGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604504 | ||||||||||||||||||||
Literature Links: |
GPR108 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR108 - G protein-coupled receptor 108 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080452.1 | 1319 | Missense Mutation | GCC,TCC | A,S 439 | NP_001073921.1 | |
XM_006722801.3 | 1319 | Missense Mutation | GCC,TCC | A,S 197 | XP_006722864.1 | |
XM_011528137.2 | 1319 | Missense Mutation | GCC,TCC | A,S 197 | XP_011526439.1 | |
XM_017027012.1 | 1319 | Missense Mutation | GCC,TCC | A,S 439 | XP_016882501.1 | |
XM_017027013.1 | 1319 | Missense Mutation | GCC,TCC | A,S 439 | XP_016882502.1 | |
XM_017027014.1 | 1319 | Missense Mutation | GCC,TCC | A,S 439 | XP_016882503.1 | |
XM_017027015.1 | 1319 | Missense Mutation | GCC,TCC | A,S 197 | XP_016882504.1 |
MIR6791 - microRNA 6791 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIP10 - thyroid hormone receptor interactor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |