Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TCCCTGGAGCTGATCTTGGCCCTAT[C/T]GCCTATGTGTGAGTCCCCTAGGGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF619 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF619 - zinc finger protein 619 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145082.2 | 597 | Missense Mutation | TCG,TTG | S,L 97 | NP_001138554.1 | |
NM_001145083.1 | 597 | Intron | NP_001138555.1 | |||
NM_001145093.2 | 597 | Intron | NP_001138565.1 | |||
XM_006713112.2 | 597 | Intron | XP_006713175.1 | |||
XM_006713113.3 | 597 | Intron | XP_006713176.1 | |||
XM_011533607.1 | 597 | Missense Mutation | TCG,TTG | S,L 97 | XP_011531909.1 | |
XM_011533608.1 | 597 | Intron | XP_011531910.1 | |||
XM_017006225.1 | 597 | Intron | XP_016861714.1 |