Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGGAGGATATGCATCCTTACAA[C/G]TATTTCTCAACCCCAGCCTTTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H11A - zinc finger CCCH-type containing 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319238.1 | Intron | NP_001306167.1 | ||||
NM_001319239.1 | Intron | NP_001306168.1 | ||||
NM_014827.5 | Intron | NP_055642.3 | ||||
XM_005245643.2 | Intron | XP_005245700.1 | ||||
XM_005245644.3 | Intron | XP_005245701.1 | ||||
XM_006711673.2 | Intron | XP_006711736.1 | ||||
XM_011510199.1 | Intron | XP_011508501.1 | ||||
XM_011510200.1 | Intron | XP_011508502.1 | ||||
XM_011510201.1 | Intron | XP_011508503.1 | ||||
XM_011510202.1 | Intron | XP_011508504.1 | ||||
XM_017002956.1 | Intron | XP_016858445.1 | ||||
XM_017002957.1 | Intron | XP_016858446.1 | ||||
XM_017002958.1 | Intron | XP_016858447.1 | ||||
XM_017002959.1 | Intron | XP_016858448.1 | ||||
XM_017002960.1 | Intron | XP_016858449.1 | ||||
XM_017002961.1 | Intron | XP_016858450.1 | ||||
XM_017002962.1 | Intron | XP_016858451.1 | ||||
XM_017002963.1 | Intron | XP_016858452.1 | ||||
XM_017002964.1 | Intron | XP_016858453.1 | ||||
XM_017002965.1 | Intron | XP_016858454.1 |