Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTGAGAGGGCATTTCCGCTCT[A/T]TTCGGCTGCGGAGCCGCAGGCTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603503 MIM: 609277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPM1 - dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOCS3 - molybdenum cofactor synthesis 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014484.4 | 891 | Missense Mutation | ATT,TTT | I,F 288 | NP_055299.1 |