Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTGAAGATGGCACCCACGAAG[G/T]TCAAGATGTAGAAGAGGAGGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126380 MIM: 603183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC1 - ERCC excision repair 1, endonuclease non-catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPM1N - protein phosphatase, Mg2+/Mn2+ dependent 1N (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTN2 - reticulon 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005619.4 | 1636 | Missense Mutation | AAC,ACC | N,T 469 | NP_005610.1 | |
NM_206900.2 | 1636 | Missense Mutation | AAC,ACC | N,T 396 | NP_996783.1 | |
NM_206901.2 | 1636 | Missense Mutation | AAC,ACC | N,T 129 | NP_996784.1 |