Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCCAGATCCTTCAGACTGTTTT[C/T]CTCTCAATCTTTGCTGTTGCTACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607579 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC22A9 - solute carrier family 22 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080866.2 | 337 | Silent Mutation | TTC,TTT | F,F 23 | NP_543142.2 | |
XM_017017159.1 | 337 | Silent Mutation | TTC,TTT | F,F 23 | XP_016872648.1 | |
XM_017017160.1 | 337 | UTR 5 | XP_016872649.1 |