Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GGAGACAGCTTTGCGGAAGGCCTCC[C/T]GCTGCAGGGTTCTCTCCTCACGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DENND2A - DENN domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318052.1 | 3291 | Missense Mutation | NP_001304981.1 | |||
NM_001318053.1 | 3291 | Intron | NP_001304982.1 | |||
NM_015689.4 | 3291 | Missense Mutation | NP_056504.3 | |||
XM_005249977.3 | 3291 | Missense Mutation | XP_005250034.1 | |||
XM_005249979.4 | 3291 | Intron | XP_005250036.1 | |||
XM_011516052.2 | 3291 | Missense Mutation | XP_011514354.1 | |||
XM_011516053.2 | 3291 | Missense Mutation | XP_011514355.1 | |||
XM_011516054.2 | 3291 | Missense Mutation | XP_011514356.1 | |||
XM_017011989.1 | 3291 | Missense Mutation | XP_016867478.1 |