Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| HIP1 - huntingtin interacting protein 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001243198.2 | 7895 | UTR 3 | NP_001230127.1 | |||
| NM_005338.6 | 7895 | UTR 3 | NP_005329.3 | |||
| XM_005250304.2 | 7895 | UTR 3 | XP_005250361.1 | |||
| XM_005250305.2 | 7895 | UTR 3 | XP_005250362.1 | |||
| XM_011516116.2 | 7895 | UTR 3 | XP_011514418.1 | |||
| XM_017012099.1 | 7895 | UTR 3 | XP_016867588.1 | |||
| PMS2P3 - PMS1 homolog 2, mismatch repair system component pseudogene 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||