Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGTGCCATCTTTCACCTCACCA[A/G]TATCAGGTATAGCCATCCCTAAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603712 | ||||||||||||||||||||
Literature Links: |
ADAM20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM20 - ADAM metallopeptidase domain 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003814.4 | 11105 | Missense Mutation | ACT,ATT | T,I 655 | NP_003805.3 | |
XM_005268151.3 | 11105 | Missense Mutation | ACT,ATT | T,I 655 | XP_005268208.1 |