Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TAGGCAAATTCTTCTGTATGAGCTG[G/T]ATCAGTAAACACGCCAATAAAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606934 MIM: 612818 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFAF1 - NADH:ubiquinone oxidoreductase complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016013.3 | 1247 | Missense Mutation | ACA,CCA | T,P 307 | NP_057097.2 | |
XM_006720555.2 | 1247 | Missense Mutation | ACA,CCA | T,P 307 | XP_006720618.1 | |
XM_011521658.1 | 1247 | Missense Mutation | ACA,CCA | T,P 307 | XP_011519960.1 | |
XM_011521659.2 | 1247 | UTR 3 | XP_011519961.1 | |||
XM_017022293.1 | 1247 | Intron | XP_016877782.1 |
NUSAP1 - nucleolar and spindle associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |