Search
Search

ALPK3CGGGAGCCGGGTCCCGGGAGCTCCA[C/T]GAAGGGGCCTGTCCTCCATGACCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ALPK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ALPK3 - alpha kinase 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020778.4 | 409 | Missense Mutation | ACG,ATG | T,M 81 | NP_065829.3 | |
| XM_017022444.1 | 409 | Missense Mutation | ACG,ATG | T,M 81 | XP_016877933.1 | |
| XM_017022445.1 | 409 | Missense Mutation | ACG,ATG | T,M 81 | XP_016877934.1 | |