Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGCTTGTGCAGAATCTTCTCGCCCC[G/C]TGACATAAATTTCACAGGGAATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607974 | ||||||||||||||||||||
Literature Links: |
NETO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NETO2 - neuropilin and tolloid like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201477.1 | 1924 | Missense Mutation | ACG,AGG | T,R 504 | NP_001188406.1 | |
NM_018092.4 | 1924 | Missense Mutation | ACG,AGG | T,R 511 | NP_060562.3 | |
XM_006721289.1 | 1924 | Missense Mutation | ACG,AGG | T,R 473 | XP_006721352.1 | |
XM_006721291.1 | 1924 | Missense Mutation | ACG,AGG | T,R 466 | XP_006721354.1 | |
XM_006721292.1 | 1924 | Intron | XP_006721355.1 | |||
XM_017023740.1 | 1924 | Missense Mutation | ACG,AGG | T,R 375 | XP_016879229.1 |