Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCGAAGACGAGGGCCTGGAGG[A/G]AACAGGTATGACTCCACTTAGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608954 | ||||||||||||||||||||
Literature Links: |
MON1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MON1B - MON1 homolog B, secretory trafficking associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286639.1 | 493 | Missense Mutation | GAA,GGA | E,G 48 | NP_001273568.1 | |
NM_001286640.1 | 493 | Intron | NP_001273569.1 | |||
NM_014940.3 | 493 | Missense Mutation | GAA,GGA | E,G 48 | NP_055755.1 |
SYCE1L - synaptonemal complex central element protein 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |