Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GCTCCAGGTGGTTGAAGGCCTCCCA[A/G]GGGCAGATGACTGTGATGTCTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180073 | ||||||||||||||||||||
Literature Links: |
PDE6G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002602.3 | 351 | Silent Mutation | CCC,CCT | P,P 69 | NP_002593.1 | |
XM_006721942.3 | 351 | Silent Mutation | CCC,CCT | P,P 119 | XP_006722005.1 | |
XM_017024734.1 | 351 | Silent Mutation | CCC,CCT | P,P 139 | XP_016880223.1 | |
XM_017024735.1 | 351 | Silent Mutation | CCC,CCT | P,P 69 | XP_016880224.1 | |
XM_017024736.1 | 351 | Silent Mutation | CCC,CCT | P,P 69 | XP_016880225.1 |
TSPAN10 - tetraspanin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |