Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CGGCCCATTTTACTTCCTGTTCCAG[A/G]CAGCCTGACCCTGGATTCTGGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152391 MIM: 608992 MIM: 610718 MIM: 615672 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALOX12 - arachidonate 12-lipoxygenase, 12S type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ALOX12-AS1 - ALOX12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BCL6B - B-cell CLL/lymphoma 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C17orf49 - chromosome 17 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142798.2 | 636 | Missense Mutation | GAC,GGC | D,G 172 | NP_001136270.1 | |
NM_001142799.2 | 636 | Intron | NP_001136271.1 | |||
NM_174893.3 | 636 | Intron | NP_777553.1 |
MIR195 - microRNA 195 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR497 - microRNA 497 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR497HG - mir-497-195 cluster host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNASEK - ribonuclease K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNASEK-C17orf49 - RNASEK-C17orf49 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |