Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_190365219_10
          See other KRTAP3-1 GT Assays ›
          SNP ID:
          rs201729934
          Gene
          KRTAP3-1 KRTAP3-2 KRTAP3-3
          Gene Name
          keratin associated protein 3-1
          keratin associated protein 3-2
          keratin associated protein 3-3
          Set Membership:
          -
          Chromosome Location:
          Chr.17: 40999852 - 40999852 on Build GRCh38
          Polymorphism:
          A/G, Transition substitution
          Context Sequence [VIC/FAM]:

          GCAGCTGCGAGAGGCACAGCAATCC[A/G]TGGCAATGGGCGTCTGGTTAGCTTT

          Assay ID C_190365219_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          Literature Links:

          KRTAP3-1 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          G (0.00)
          (1.00)
          Caucasian - Not Available CEPH (CEU) - Not Available
          EAS
          G (0.00)
          (1.00)
          African American - Not Available YRI (Yoruba) - Not Available
          SAS
          G (0.00)
          (1.00)
          Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR
          G (0.00)
          (1.00)
          Japanese - Not Available JPT (Japanese) - Not Available
          EUR
          G (0.00)
          (1.00)
          AMR
          G (0.00)
          (1.00)
          KRTAP3-1 - keratin associated protein 3-1
          There are no transcripts associated with this gene.
          KRTAP3-2 - keratin associated protein 3-2
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_031959.2 35 Missense Mutation ACG,ATG T,M 1 NP_114165.1
          KRTAP3-3 - keratin associated protein 3-3
          There are no transcripts associated with this gene.

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          structural molecule activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline