Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
CGTGTTGATCTCACACTCCAGCCGG[G/T]CCCGGACGTCCAGCAGCACCTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602760 MIM: 602764 MIM: 604540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT32 - keratin 32 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT35 - keratin 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002280.4 | 2809 | Missense Mutation | NP_002271.3 | |||
XM_011524794.1 | 2809 | Missense Mutation | XP_011523096.1 |
KRT36 - keratin 36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |