Search Thermo Fisher Scientific
CTGTCCTGCCAGCTGTACCAGAGAT[C/T]GGGAGACATGGGCCTCGGTGTGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607427 MIM: 188350 | ||||||||||||||||||||
Literature Links: |
ENOSF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENOSF1 - enolase superfamily member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126123.3 | 4930 | UTR 3 | NP_001119595.1 | |||
NM_001318759.1 | 4930 | UTR 3 | NP_001305688.1 | |||
NM_001318760.1 | 4930 | Intron | NP_001305689.1 | |||
NM_017512.5 | 4930 | UTR 3 | NP_059982.2 | |||
NM_202758.3 | 4930 | UTR 3 | NP_974487.1 | |||
XM_011525679.2 | 4930 | Intron | XP_011523981.2 | |||
XM_017025833.1 | 4930 | Intron | XP_016881322.1 | |||
XM_017025834.1 | 4930 | Intron | XP_016881323.1 | |||
XM_017025835.1 | 4930 | Intron | XP_016881324.1 | |||
XM_017025836.1 | 4930 | Intron | XP_016881325.1 | |||
XM_017025837.1 | 4930 | Intron | XP_016881326.1 |
TYMS - thymidylate synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001071.2 | 4930 | Missense Mutation | TCG,TTG | S,L 216 | NP_001062.1 |