Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAACCCTGCTCAGAGGAATTTG[C/T]ACAGGAAAGTGATGTTGGAGAACTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF583 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF583 - zinc finger protein 583 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159860.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | NP_001153332.1 | |
NM_001159861.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | NP_001153333.1 | |
NM_152478.2 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | NP_689691.2 | |
XM_005258558.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 32 | XP_005258615.1 | |
XM_011526517.2 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | XP_011524819.1 | |
XM_017026346.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016881835.1 | |
XM_017026347.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016881836.1 | |
XM_017026348.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016881837.1 | |
XM_017026349.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016881838.1 | |
XM_017026350.1 | 2092 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016881839.1 | |
XM_017026351.1 | 2092 | UTR 5 | XP_016881840.1 | |||
XM_017026352.1 | 2092 | UTR 5 | XP_016881841.1 | |||
XM_017026353.1 | 2092 | UTR 5 | XP_016881842.1 |