Search
Search

ATP1A3
RABAC1TGGCTGGGGCGGGAGGAATGGATAG[A/G]GGGGTGAGGAGAGGGAGAGAAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ATP1A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ATP1A3 - ATPase Na+/K+ transporting subunit alpha 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001256213.1 | 3517 | UTR 3 | NP_001243142.1 | |||
| NM_001256214.1 | 3517 | UTR 3 | NP_001243143.1 | |||
| NM_152296.4 | 3517 | UTR 3 | NP_689509.1 | |||
| RABAC1 - Rab acceptor 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||